Learn More
Selleck Chemical LLC PIK-93 S1489-1g
SDP
Catalog No. 503111320
Encompass_Preferred
PIK-93 is the first potent synthetic PI4K (PI4KIII ) inhibitor with IC50 of 19 nM shown to inhibit PI3K with IC50 of 39 nM
Catalog No. 50-311-1320 Supplier Selleck Chemical LLC Supplier No. S14891G
Add to Cart
Add to Cart
PIK-93 is the first potent synthetic PI4K (PI4KIII ) inhibitor with IC50 of 19 nM shown to inhibit PI3K with IC50 of 39 nM