Learn More
Echelon Biosciences Research Labs PIK-93 5 mg
SDP
Catalog No. 5082262
Encompass
PIK-93 is the first reported PI4-kinase inhibitor, which is able to inhibit PI4KIII at low-nanomolar range (IC50=19 nM). In addition, this compound is a potent inhibitor of PI3K in vitro (IC50=16 nM). This compound inhibits p110, p110, p110 with IC50s of 0.039, 0.59, 0.12 M respectively. PIK-93 hydrogen bonds to the backbone amide and carbonyl of Val882 and between its sulphonamide and Asp964. PI4KIII is esential for replication of numerous viruses P. falciparum. Inhibition of plasmodial PI4KIII has been proposed as an anti-malaria treatment. This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-822-62 Supplier Echelon Biosciences Research Labs Supplier No. B03065MG
Add to Cart
Add to Cart
PIK-93 is the first reported PI4-kinase inhibitor, which is able to inhibit PI4KIII at low-nanomolar range (IC50=19 nM). In addition, this compound is a potent inhibitor of PI3K in vitro (IC50=16 nM). This compound inhibits p110, p110, p110 with IC50s of 0.039, 0.59, 0.12 M respectively. PIK-93 hydrogen bonds to the backbone amide and carbonyl of Val882 and between its sulphonamide and Asp964. PI4KIII is esential for replication of numerous viruses P. falciparum. Inhibition of plasmodial PI4KIII has been proposed as an anti-malaria treatment. This item is non returnable