Learn More
Selleck Chemical LLC PIK-III
SDP
Catalog No. 5000000306
Encompass_Preferred
PIK-III (VPS34-IN2) which is a selective inhibitor of VPS34 enzymatic activity inhibits autophagy and de novo lipidation of LC3 and leads to the stabilization of autophagy substrates The IC50 values for VPS34 and PI(3)K are 0 018 M and 1 2 M respectively
Catalog No. 50-000-00306 Supplier Selleck Chemical LLC Supplier No. S768325MG
PIK-III (VPS34-IN2) which is a selective inhibitor of VPS34 enzymatic activity inhibits autophagy and de novo lipidation of LC3 and leads to the stabilization of autophagy substrates The IC50 values for VPS34 and PI(3)K are 0 018 M and 1 2 M respectively