Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human PIK3CA E545K/PIK3R1 (p110 alpha E545K/p85 alpha), His Tag Recombinant Protein

Catalog No. A31083
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A31083 20 μg
A32909 100 μg
A33394 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A31083 Supplier Invitrogen™ Supplier No. A31083
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This PIK3CA E545K/PIK3R1 (p110 alpha E545K/p85 alpha) recombinant human protein (full length) was expressed in insect cells. PIK3CA E545K/PIK3R1 (p110 alpha E545K/p85 alpha) recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the Adapta™ assay Available in multiple pack sizes for convenient use Also available in our SelectScreen Kinase Profiling Service Protein Form: Full Length, Recombinant, Active, Mutant The gene accession number for PIK3CA (p110α) is NP_006209.2 and for PIK3R1 (p85α) is NP_852664.1.

PIK3CA E542K/PIK3R1 (p110 alpha E542K/p85 alpha) lipid kinase is comprised of a 110 kDa catalytic subunit and an 85 kDa regulatory subunit. A number of isoforms of the 110 kDa catalytic subunit and the 85 kDa regulatory subunit exist in cells. PIK3CA is frequently mutated or amplified in a variety of cancers including ovarian and colon and this kinase is one of the PI3K mutants. PIK3CA gene copy number is increased in over 30% of ovarian cancers and this leads to increased PI3-kinase activity. Furthermore, the activity of PIK3CA is essential for vascular development and inactivation of PIK3CA leads to severe defects in angiogenic sprouting and vascular remodeling.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P27986, P42336
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM sodium phosphate with 150mM Imidazole, 0.1mM PMSF, 0.25mM DTT, 25% glycerol, 300mM NaCl and no preservative; pH 7
Gene ID (Entrez) 5290, 5295
Molecular Weight (g/mol) 125.4-84.7 kDa
Name Human PIK3CA E545K/PIK3R1 (p110 alpha E545K/p85 alpha), His Tag
pH Range 7
Purification Method Purified
Quantity 20 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias PIK3CA; PIK3R1
Common Name PIK3CA (E545K Mutant)/PIK3R1
Gene Symbol PIK3CA, PIK3R1
Kinase Family PI3 Kinase
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Shipping Condition Dry Ice
Expression System Insect cells
Protein Family Kinases & Inhibitors
Protein Form Full Length, Recombinant, Active, Mutant
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.