Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human PIK3CA/PIK3R1 (p110 alpha/p85 alpha), His Tag Recombinant Protein

Catalog No. PV4788
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV4788 20 μg
PR8581C 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PV4788 Supplier Invitrogen™ Supplier No. PV4788
Add to Cart
Edge
Add to Cart

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length.

PIK3CA (p110 alpha) is known to be involved in the PI 3-kinase/AKT signaling pathway, which plays an important role in regulating cell growth and apoptosis. PIK3CA is coexpressed and copurified with the regulatory subunit PI3KR1.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P27986, P42336
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris HCl with 150mM NaCl, 50% glycerol, 0.5mM EDTA, 2mM DTT, 0.04% Triton X-100 and no preservative; pH 7.5
Gene ID (Entrez) 5290, 5295
Molecular Weight (g/mol) 212 kDa
Name Human PIK3CA/PIK3R1 (p110 alpha/p85 alpha), His Tag
pH Range 7.5
Purification Method Purified
Quantity 20 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias PIK3CA; PIK3R1
Common Name PIK3CA/PIK3R1
Gene Symbol PIK3CA, PIK3R1
Kinase Family PI3 Kinase Family
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Assay Entry Biochemical binding/kinetic, Biochemical ADP accumulation
Kinase Group Lipid Kinases
Shipping Condition Dry Ice
System Compatibility Adapta™
Tag Position His (N-Terminal)⁄None
Target Entry PIK3CA/PIK3R1
Target Gene phosphoinositide-3-kinase, catalytic, alpha polypeptide
Expression System Baculovirus
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Full Length
Form Liquid
Protein Subtype Lipid Kinases
Research Category Signal Transduction
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.