Learn More
Aviva Systems Biology Corp PIK3CB Antibody - C-terminal region (ARP32117_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32117_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PIK3CB. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human PIK3CB. Alternate names: PI3K, PIK3C1, P110BETA, PI3KBETAApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54454