Learn More
Aviva Systems Biology Corp PIK3R5 Antibody - middle region (ARP55012_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55012_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PIK3R5. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human PIK3R5. Alternate names: F730038I15Rik, FOAP-2, P101-PI3K, p101ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-990