Learn More
Cayman Chemical PIK75 hydrochloride
SDP
Catalog No. 502890303
Encompass_Preferred
A selective inhibitor of p110α with an IC50 value of 5.8 nM; inhibits p110γ and p110β with IC50 values of 0.076 µM and 1.3 µM, respectively
Catalog No. 50-289-0303 Supplier Cayman Chemical Supplier No. 1000921010 MG
A selective inhibitor of p110α with an IC50 value of 5.8 nM; inhibits p110γ and p110β with IC50 values of 0.076 µM and 1.3 µM, respectively