Learn More
Cayman Chemical PIK-75, Purity: ≥98%, CAS: 372196-67-3, 25 mg
SDP
Catalog No. 502890870
Encompass_Preferred
A selective inhibitor of p110α with an IC50 value of 5.8 nM; inhibits p110γ and p110β with IC50 values of 0.076 µM and 1.3 µM, respectively
Catalog No. 50-289-0870 Supplier Cayman Chemical Supplier No. 1000921025 MG
A selective inhibitor of p110α with an IC50 value of 5.8 nM; inhibits p110γ and p110β with IC50 values of 0.076 µM and 1.3 µM, respectively