Learn More
RESONON INC Pika L Hyperspectral Benchtop System contains all you need to capture and analyze hyperspectral data across the 400-1000nm spectrum. Includes imager, lens, and stage.
SDP
Catalog No. NC2953195
Encompass_Preferred
Resonon's Pika L Hyperspectral Benchtop System contains all you need to capture and analyze hyperspectral data across the 400-1000nm spectrum. It includes a line-scan imager, and a Benchtop Reflectance kit featuring a translation stage, halogen lighting, and a data acquistion system. Spectronon software provides data visualization, multiple analysis tools, and accomodates user-written plugins. The Pika L is Resonon's most popular hyperspectral camera.
Catalog No. NC2953195 Supplier RESONON INC Supplier No. RES0001
Add to Cart
Add to Cart
Request a Quote
Resonon's Pika L Hyperspectral Benchtop System contains all you need to capture and analyze hyperspectral data across the 400-1000nm spectrum. It includes a line-scan imager, and a Benchtop Reflectance kit featuring a translation stage, halogen lighting, and a data acquistion system. Spectronon software provides data visualization, multiple analysis tools, and accomodates user-written plugins. The Pika L is Resonon's most popular hyperspectral camera.