Learn More
RESONON INC Pika XC2 Hyperspectral Benchtop System contains all you need to capture and analyze high resulution hyperspectral data across the 400-1000nm spectrum. Includes imager, lens, and stage.
SDP
Catalog No. NC2953202
Encompass_Preferred
Resonon's Pika XC2 Hyperspectral Benchtop System contains all you need to capture and analyze high resolution hyperspectral data across the 400-1000nm spectrum.It includes a line-scan imager, and a Benchtop Reflectance kit featuring a translation stage, halogen lighting, and a data acquistion system. Spectronon software provides data visualization, multiple analysis tools, and accomodates user-written plugins. The Pika XC2 has best in-class spectral resolution, and excellent imaging quality.
Catalog No. NC2953202 Supplier RESONON INC Supplier No. RES0002
Add to Cart
Add to Cart
Request a Quote
Resonon's Pika XC2 Hyperspectral Benchtop System contains all you need to capture and analyze high resolution hyperspectral data across the 400-1000nm spectrum.It includes a line-scan imager, and a Benchtop Reflectance kit featuring a translation stage, halogen lighting, and a data acquistion system. Spectronon software provides data visualization, multiple analysis tools, and accomodates user-written plugins. The Pika XC2 has best in-class spectral resolution, and excellent imaging quality.