Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Piketoprofen | 60576-13-8 | MFCD00866026 | 99.4% | 344.41 g/mol | C22H20N2O2 | 250 MG
SDP

Catalog No. 5000244002
Encompass_Preferred

Piketoprofen is a nonsteroidal anti-inflammatory research chemical used primarily in topical formulations to study soft tissue rheumatism and related inflammatory models; piketoprofen-amide is noted as a prodrug of a COX-2 inhibitor. It is supplied as a solid and as a DMSO solution, with supporting SDS and product data sheet documentation.

  • Nonsteroidal anti-inflammatory reagent for topical and soft tissue inflammation studies.
  • Available as solid and as a 10 mM DMSO solution; multiple mass sizes available, including 250 mg.
  • High purity (≈99.4%) suitable for research and analytical applications.
  • Molecular formula C22H20N2O2; molecular weight 344.41 g/mol.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • SDS and product data sheet available for handling and safety information.
  • For research use only; not for human or clinical use.

Catalog No. 50-002-44002 Supplier Medchemexpress LLC Supplier No. HY108297250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Piketoprofen is a nonsteroidal anti-inflammatory research chemical used primarily in topical formulations to study soft tissue rheumatism and related inflammatory models; piketoprofen-amide is noted as a prodrug of a COX-2 inhibitor. It is supplied as a solid and as a DMSO solution, with supporting SDS and product data sheet documentation.

  • Nonsteroidal anti-inflammatory reagent for topical and soft tissue inflammation studies.
  • Available as solid and as a 10 mM DMSO solution; multiple mass sizes available, including 250 mg.
  • High purity (≈99.4%) suitable for research and analytical applications.
  • Molecular formula C22H20N2O2; molecular weight 344.41 g/mol.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • SDS and product data sheet available for handling and safety information.
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.