Learn More
GA International PikPAD™, Application Pad, 6" x 4", purple (Pack of 3) 3 applicator/pack
Catalog No. 5000561991
Encompass_Preferred
GA International’s PikPad™ facilitates the application of small dot labels when used with the PikaTAG applicator. PikPad is a soft, reusable, and durable pad that acts as a flat surface to maximize the number of labels that can be picked using the PikaTAG applicator in a given time, significantly improving label picking efficiency. Simply place the PikaPad underneath the label liner and reliably pick up dot labels in a single attempt*. They are designed to work with the PikaTAG applicator and all PikaTAG-compatible labels, saving you precious time and resulting in near-effortless label placement.
Catalog No. 50-005-61991 Supplier GA International Supplier No. PIKPAD13
GA International’s PikPad™ facilitates the application of small dot labels when used with the PikaTAG applicator. PikPad is a soft, reusable, and durable pad that acts as a flat surface to maximize the number of labels that can be picked using the PikaTAG applicator in a given time, significantly improving label picking efficiency. Simply place the PikaPad underneath the label liner and reliably pick up dot labels in a single attempt*. They are designed to work with the PikaTAG applicator and all PikaTAG-compatible labels, saving you precious time and resulting in near-effortless label placement.