Learn More
Cayman Chemical Pikromycin, Purity: ≥95%, CAS: 19721-56-3, 2.5 mg
SDP
Catalog No. 502990747
Encompass_Preferred
A macrolide antibiotic; active against E. coli, S. aureus, and B. subtilis (MIC99s = 50-60, 90-100, and 25-30 µM, respectively)
Catalog No. 50-299-0747 Supplier Cayman Chemical Supplier No. 313312.5 MG
A macrolide antibiotic; active against E. coli, S. aureus, and B. subtilis (MIC99s = 50-60, 90-100, and 25-30 µM, respectively)