Learn More
AdipoGen Pikromycin
SDP
Supplier:  AdipoGen BVT0400M005
Encompass_Preferred
Chemical. CAS 19721-56-3. Formula C28H47NO8. MW 525.7. Isolated from Streptomyces sp. Antibiotic. 14-Membered macrolide. Active against Gram-positive bacteria including mycobacteria. Prolyl endopeptidase PREP inhibitor.
Catalog No. 50-256-1947