Learn More
Medchemexpress LLC Pilabactam sodium | 2410688-61-6 | 99.5% | 262.19 | 1 ML
SDP
Catalog No. 5000359994
Encompass_Preferred
Pilabactam sodium (ANT3310) is a broad-spectrum covalent Serine β-Lactamase inhibitor with IC50 values ranging from 1 nM to 175 nM across a panel of Serine β-Lactamase. It potentiates the activity of β-lactam antibiotics against Carbapenem-Resistant Enterobacterales (CRE) and Acinetobacter baumannii (CRAB), making it valuable for bacterial infection research.
  • Broad-spectrum covalent Serine β-Lactamase inhibitor
  • Potentiates activity of β-lactam antibiotics
  • Effective against Carbapenem-Resistant Enterobacterales (CRE)
  • Effective against Acinetobacter baumannii (CRAB)
  • Low in vitro cytotoxicity (>100 μM)
  • Low cardiotoxicity
  • No genotoxicity
Catalog No. 50-003-59994 Supplier Medchemexpress LLC Supplier No. HY14734910MM/1ML
Pilabactam sodium (ANT3310) is a broad-spectrum covalent Serine β-Lactamase inhibitor with IC50 values ranging from 1 nM to 175 nM across a panel of Serine β-Lactamase. It potentiates the activity of β-lactam antibiotics against Carbapenem-Resistant Enterobacterales (CRE) and Acinetobacter baumannii (CRAB), making it valuable for bacterial infection research.
  • Broad-spectrum covalent Serine β-Lactamase inhibitor
  • Potentiates activity of β-lactam antibiotics
  • Effective against Carbapenem-Resistant Enterobacterales (CRE)
  • Effective against Acinetobacter baumannii (CRAB)
  • Low in vitro cytotoxicity (>100 μM)
  • Low cardiotoxicity
  • No genotoxicity