Learn More
Medchemexpress LLC HY-11105 10mg Medchemexpress, Pilaralisib analogue CAS:956958-53-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1110510MG
Encompass_Preferred
Medchemexpress, HY-11105 10mg Pilaralisib analogue CAS:956958-53-5 Pilaralisib analogue (XL147 analogue) is a representative and selective PI3Kα inhibitor extracted from patent WO2012006552A1, Compound 147 in Table 1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0787