Learn More
Cayman Chemical PIlIformIc AcId 2.5mg
SDP
Catalog No. 502992245
Encompass_Preferred
A fungal metabolite with diverse biological activities; cytotoxic to BC-1 human breast cancer cells (IC50 = 5 UG/ml); active against L. braziliensis amastigotes (IC50 = 78.5 µM); active against the plant pathogenic fungi C. gloeosporioides (MIC = 292 µM)
Catalog No. 50-299-2245 Supplier Cayman Chemical Supplier No. 317562.5 MG
A fungal metabolite with diverse biological activities; cytotoxic to BC-1 human breast cancer cells (IC50 = 5 UG/ml); active against L. braziliensis amastigotes (IC50 = 78.5 µM); active against the plant pathogenic fungi C. gloeosporioides (MIC = 292 µM)