Learn More
Fisherbrand™ Pill Counting Tray Triangular, Pack of 10
Catalog No. 14127543 Shop All Fisherbrand Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility
14-127-543 Non-sterile
14-127-539 Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-127-543
Add to Cart
Add to Cart
Streamlined for safe, fast distribution
  • Disposable to ensure no chemical cross-contamination for patient safety
  • Made from FDA food grade polystyrene in an ISO Class 8 clean room
  • Meets quality control and GMP requirements for manufacturing batch sampling
  • Durable and shatterproof trays constructed of inert BSE/TSE free material
  • Lot traceable
  • Pour spout in corner for dispensing control
  • Ergonomic spatula length - 6.9 in./17.6 cm
  • Available in both sterile and non-sterile for intended usage
Specifications
Dimensions (L x W x H) 9.1 x 9.1 x 0.5 in.
Material Polystyrene
Type Tray
Quantity 10/Bag
Shape Triangular
Sterility Non-sterile