Learn More
VWR INTERNATIONAL LLC PILLOW BAG 10L 10/CS
Catalog No. NC2263565
Encompass
Pillow bag, 3 ports welding; C-Flex 10 L Tube plug, ³/₈" Int.Ø × 5/8" Ext.Ø; length: 11.75" Tube plug, ³/₈ Int.Ø × 5/8" Ext.Ø; length: 11.75" Female luer and needleless swabable valve and cap; P3: ¹/₄ Int.Ø × ⁷/₁₆" Ext.Ø; length: 2"
This item is not returnable. View return policy
Catalog No. NC2263565 Supplier VWR INTERNATIONAL LLC Supplier No. 76358688
Pillow bag, 3 ports welding; C-Flex 10 L Tube plug, ³/₈" Int.Ø × 5/8" Ext.Ø; length: 11.75" Tube plug, ³/₈ Int.Ø × 5/8" Ext.Ø; length: 11.75" Female luer and needleless swabable valve and cap; P3: ¹/₄ Int.Ø × ⁷/₁₆" Ext.Ø; length: 2"