Learn More
Apexbio Technology LLC Pilocarpine HCl(Synonyms: Pilocarpine hydrochloride, Salagen, Pilocar, Isopto Carpine), 10mM (in 1mL DMSO), CAS: 54-71-7.
SDP
Catalog No. 5000060062
Encompass_Preferred
Pilocarpine HCl (CAS 54-71-7) the hydrochloride salt form of pilocarpine is a nonselective muscarinic acetylcholine receptor agonist derived from plants of the Pilocarpus genus By directly activating muscarinic receptors pilocarpine promotes exocrine gland secretion induces contraction of smooth muscle tissues and when applied ocularly triggers miosis and ciliary muscle contraction enhancing aqueous humor drainage and reducing intraocular pressure In biomedical research pilocarpine is employed widely to induce rodent epilepsy models and to study cholinergic mechanisms underlying secretory processes
Catalog No. 50-000-60062 Supplier Apexbio Technology LLC Supplier No. B18165.1
Pilocarpine HCl (CAS 54-71-7) the hydrochloride salt form of pilocarpine is a nonselective muscarinic acetylcholine receptor agonist derived from plants of the Pilocarpus genus By directly activating muscarinic receptors pilocarpine promotes exocrine gland secretion induces contraction of smooth muscle tissues and when applied ocularly triggers miosis and ciliary muscle contraction enhancing aqueous humor drainage and reducing intraocular pressure In biomedical research pilocarpine is employed widely to induce rodent epilepsy models and to study cholinergic mechanisms underlying secretory processes