Learn More
Selleck Chemical LLC Pilocarpine HCl 50mg
SDP
Catalog No. 502173139
Encompass_Preferred
Pilocarpine HCl (NSC 5746) is a nonselective muscarinic acetylcholine receptor agonist used to produce an experimental model of epilepsy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3139 Supplier Selleck Chemical LLC Supplier No. S423150MG
Pilocarpine HCl (NSC 5746) is a nonselective muscarinic acetylcholine receptor agonist used to produce an experimental model of epilepsy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.