Learn More
Medchemexpress LLC Pilocarpine (hydrochloride) | 54-71-7 | 99.92% | 1 G
SDP
Catalog No. 5000408189
Encompass_Preferred
Pilocarpine Hydrochloride is a potent M3-type muscarinic acetylcholine receptor (M3 muscarinic receptor) agonist, identified for use as a laboratory chemical and in the manufacture of substances.
  • Acts as an agonist for M3-type muscarinic acetylcholine receptors
  • Relevant for research in cardiovascular disease, neurological disease, and cancer
  • Has a purity of 99.92% as determined by LCMS
  • Appears as a white to off-white solid
  • Recommended storage at 4°C, sealed, and away from moisture
  • Unstable in solutions, fresh preparation is recommended
Catalog No. 50-004-08189 Supplier Medchemexpress LLC Supplier No. HYB07261G
Pilocarpine Hydrochloride is a potent M3-type muscarinic acetylcholine receptor (M3 muscarinic receptor) agonist, identified for use as a laboratory chemical and in the manufacture of substances.
  • Acts as an agonist for M3-type muscarinic acetylcholine receptors
  • Relevant for research in cardiovascular disease, neurological disease, and cancer
  • Has a purity of 99.92% as determined by LCMS
  • Appears as a white to off-white solid
  • Recommended storage at 4°C, sealed, and away from moisture
  • Unstable in solutions, fresh preparation is recommended