Learn More
Medchemexpress LLC Pilocarpine hydrochloride | 54-71-7 | 99.98% | 5 G
SDP
Catalog No. 5000406756
Encompass_Preferred
Pilocarpine Hydrochloride is a potent M3-type muscarinic acetylcholine receptor (M3 muscarinic receptor) agonist. It is intended for research use only.
  • Potent M3-type muscarinic acetylcholine receptor agonist
  • Unstable in solutions, freshly prepared is recommended
  • Suitable for laboratory centrifugation and storage
Catalog No. 50-004-06756 Supplier Medchemexpress LLC Supplier No. HYB07265G
Pilocarpine Hydrochloride is a potent M3-type muscarinic acetylcholine receptor (M3 muscarinic receptor) agonist. It is intended for research use only.
  • Potent M3-type muscarinic acetylcholine receptor agonist
  • Unstable in solutions, freshly prepared is recommended
  • Suitable for laboratory centrifugation and storage