Learn More
Medchemexpress LLC HY-B0726 500mg Medchemexpress, Pilocarpine (Hydrochloride) CAS:54-71-7 Purity:98%
SDP
Catalog No. 501639592
Encompass_Preferred
Medchemexpress,HY-B0726 500mg Pilocarpine (Hydrochloride) CAS:54-71-7 Formula:C11H17ClN2O2 Pilocarpine Hydrochloride is a selective M3-type muscarinic acetylcholine receptor (M3 muscarinic receptor) agonist. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-9592 Supplier Medchemexpress LLC Supplier No. HYB0726500MG
Medchemexpress,HY-B0726 500mg Pilocarpine (Hydrochloride) CAS:54-71-7 Formula:C11H17ClN2O2 Pilocarpine Hydrochloride is a selective M3-type muscarinic acetylcholine receptor (M3 muscarinic receptor) agonist. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.