Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pilocarpine hydrochloride, 99.3%, MP Biomedicals™

Catalog No. ICN15189201 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15189201 1 g
ICN15189205 5 g
ICN15189210 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15189201 Supplier MP Biomedicals, Inc Supplier No. 0215189201
Add to Cart
Add to Cart

Chemical Identifiers

CAS 54-71-7
Molecular Formula C11H17ClN2O2
Molecular Weight (g/mol) 244.72
MDL Number MFCD00012722
InChI Key RNAICSBVACLLGM-GNAZCLTHSA-N
Synonym pilocarpine hydrochloride, +-pilocarpine hydrochloride, pilocarpal, pilocar, pilocar smp, almocarpine, pilomiotin, pilovisc, pilocel, pilocarpine hcl
PubChem CID 5909
IUPAC Name hydrogen (3S,4R)-3-ethyl-4-[(1-methyl-1H-imidazol-5-yl)methyl]oxolan-2-one chloride
SMILES [H+].[Cl-].CC[C@H]1[C@@H](CC2=CN=CN2C)COC1=O

Specifications

Assay Percent Range 0.993
Quantity 1 g
Formula Weight 244.7
Physical Form Crystalline
Chemical Name or Material Pilocarpine hydrochloride
Recommended Storage Store at Room Temperature.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.