Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pilocarpine nitrate | 148-72-1 | MFCD00078497 | 99.8% | 100 MG
SDP

Catalog No. 5000407474
Encompass_Preferred

Pilocarpine nitrate is a potent M3-type muscarinic acetylcholine receptor agonist. It is identified by PubChem CID 657349, with a molecular formula of C11H17N3O5 and a molecular weight of 271.27 g/mol. Classified as a miotic and a muscarinic agonist, it is provided as a solid.

  • Potent M3-type muscarinic acetylcholine receptor agonist
  • Molecular formula C11H17N3O5
  • Molecular weight 271.27 g/mol
  • Appearance: Solid
  • Color: White to off-white
  • Storage: 4°C, sealed, away from moisture
  • Unstable in solutions; freshly prepared solutions are recommended
  • Acutely toxic if swallowed or inhaled

Catalog No. 50-004-07474 Supplier Medchemexpress LLC Supplier No. HYB1006100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pilocarpine nitrate is a potent M3-type muscarinic acetylcholine receptor agonist. It is identified by PubChem CID 657349, with a molecular formula of C11H17N3O5 and a molecular weight of 271.27 g/mol. Classified as a miotic and a muscarinic agonist, it is provided as a solid.

  • Potent M3-type muscarinic acetylcholine receptor agonist
  • Molecular formula C11H17N3O5
  • Molecular weight 271.27 g/mol
  • Appearance: Solid
  • Color: White to off-white
  • Storage: 4°C, sealed, away from moisture
  • Unstable in solutions; freshly prepared solutions are recommended
  • Acutely toxic if swallowed or inhaled

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.