Learn More
Grainger Protech Pilot Assembly, Liquid Propane, Metal, 1 ea
Catalog No. 502758500
Encompass_Preferred
This pilot assembly for liquid propane (LP) systems provides essential functionality for maintaining consistent operation in compatible heating units. It serves to connect the burner to the gas valve while facilitating the pilot flame necessary to ignite the main burner. Designed with an integrated sensor, this component includes a thermocouple to ensure safety by shutting off the gas supply if the pilot light is extinguished.
  • Provides a reliable pilot flame for main burner ignition
  • Connects the burner to the gas valve
  • Includes a thermocouple for automatic gas shut-off safety
  • Constructed from durable metal
  • Genuine factory authorized part
  • Compliant with Safe Drinking Water Act requirements for low lead
Catalog No. 50-275-8500 Supplier Grainger Supplier No. 15A605
This pilot assembly for liquid propane (LP) systems provides essential functionality for maintaining consistent operation in compatible heating units. It serves to connect the burner to the gas valve while facilitating the pilot flame necessary to ignite the main burner. Designed with an integrated sensor, this component includes a thermocouple to ensure safety by shutting off the gas supply if the pilot light is extinguished.
  • Provides a reliable pilot flame for main burner ignition
  • Connects the burner to the gas valve
  • Includes a thermocouple for automatic gas shut-off safety
  • Constructed from durable metal
  • Genuine factory authorized part
  • Compliant with Safe Drinking Water Act requirements for low lead