Learn More
Grainger Crown Verity NG Pilot Control Valve, Natural Gas, 1 ea
Catalog No. 502794435
Encompass_Preferred
This NG pilot control valve is designed to regulate natural gas flow to the pilot burner, facilitating reliable ignition and flame stability in compatible commercial or residential grills. It serves as a replacement component to maintain the safe and efficient operation of outdoor grilling equipment.
  • Operates on natural gas
  • Compatible with built-in gas grill models
  • Compatible with part numbers 12H003, MG-36, and BI-48 NG
  • Ensures proper gas flow for consistent pilot burner performance
Catalog No. 50-279-4435 Supplier Grainger Supplier No. 19YZ82
This NG pilot control valve is designed to regulate natural gas flow to the pilot burner, facilitating reliable ignition and flame stability in compatible commercial or residential grills. It serves as a replacement component to maintain the safe and efficient operation of outdoor grilling equipment.
  • Operates on natural gas
  • Compatible with built-in gas grill models
  • Compatible with part numbers 12H003, MG-36, and BI-48 NG
  • Ensures proper gas flow for consistent pilot burner performance