Learn More
Tingley Pilot G2 Plain Toe Knee Boot
Catalog No. 19819274 Shop All Tingley Rubber Co. Products
Change view
Click to view available options
Size:
4
5
6
7
8
9
10
11
12
13
14
15
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-819-274 4
19-819-275 5
19-819-276 6
19-819-277 7
19-819-278 8
19-819-279 9
19-819-280 10
19-819-281 11
19-819-282 12
19-819-283 13
19-819-284 14
19-819-285 15
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
California Proposition 65 - RBR Warning
Catalog No. 19-819-274 Supplier Tingley Supplier No. 31161.04
Only null left
Add to cart
Add to cart
California Proposition 65 - RBR Warning
Affordable, all day comfort. When you are working in wet, muddy conditions, our general purpose Pilot G2™ PVC knee boots provide easy walking and all day protection.
  • Injection molded construction for 100% waterproof protection
  • 15 in. height with a cutline at 12 in. to facilitate fast and clean height modification
  • Flexible upper material stays supple in cold temperatures to make walking easier
  • Tougher outsole material for good abrasion resistance
  • Self-cleaning cleated outsole spits out debris as you walk
  • Removable contour insole wicks perspiration to help keep feet dry and comfortable
Specifications
Color White
Material PVC
Size 4
Toe Material Plain Toe
Gender Unisex
Waterproof Yes
Product Type Knee Boot
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov