Learn More
Grainger Pilot Light Lens 30mm Plastic Yellow
Catalog No. 502750440
Encompass_Preferred
The Eaton 14A164 is a 30mm pilot light lens for indicating the status of electrical equipment. It has a clear polycarbonate lens and chrome-plated brass housing suitable for 22mm cutouts. The bulb is replaceable and it uses a T3-1/4 wedge base bulb up to 250V. This pilot light assembly is commonly used on control panels and industrial equipment. With its rugged construction, it provides long service life and high visibility. The overall dimensions are 30.5mm diameter x 35.6mm depth. It is UL/CSA recognized and meets IP65 ingress protection ratings.
Catalog No. 50-275-0440 Supplier Grainger Supplier No. 14A164
The Eaton 14A164 is a 30mm pilot light lens for indicating the status of electrical equipment. It has a clear polycarbonate lens and chrome-plated brass housing suitable for 22mm cutouts. The bulb is replaceable and it uses a T3-1/4 wedge base bulb up to 250V. This pilot light assembly is commonly used on control panels and industrial equipment. With its rugged construction, it provides long service life and high visibility. The overall dimensions are 30.5mm diameter x 35.6mm depth. It is UL/CSA recognized and meets IP65 ingress protection ratings.