Learn More
Grainger Pilot Light Lens 30mm Plastic Amber
Catalog No. 502753068
Encompass_Preferred
The Eaton 14A151 is a 30mm diameter pilot light lens for indicating the status of electrical equipment. It has a clear polycarbonate lens and chrome-plated brass housing suitable for 22mm cutout. The bulb is replaceable and it uses a T3-1/4 tubular LED rated for 125V AC/DC. This pilot light can be panel mounted via the threaded ring and provides 360° visibility. With its compact size, it's designed for applications with limited space.
Catalog No. 50-275-3068 Supplier Grainger Supplier No. 14A151
The Eaton 14A151 is a 30mm diameter pilot light lens for indicating the status of electrical equipment. It has a clear polycarbonate lens and chrome-plated brass housing suitable for 22mm cutout. The bulb is replaceable and it uses a T3-1/4 tubular LED rated for 125V AC/DC. This pilot light can be panel mounted via the threaded ring and provides 360° visibility. With its compact size, it's designed for applications with limited space.