Learn More
Grainger Pilot Light Lens 30mm Plastic Clear
Catalog No. 502752262
Encompass_Preferred
The Eaton 14A159 is a 30mm pilot light lens for indicating the status of electrical equipment. It has a clear polycarbonate lens and chrome-plated brass housing suitable for 22mm cutouts. The bulb is replaceable and it uses a T3-1/4 wedge base bulb up to 250V. This pilot light assembly is commonly used on control panels and industrial equipment. With its durable construction, it provides long-lasting illumination to visually indicate power or status.
Catalog No. 50-275-2262 Supplier Grainger Supplier No. 14A153
The Eaton 14A159 is a 30mm pilot light lens for indicating the status of electrical equipment. It has a clear polycarbonate lens and chrome-plated brass housing suitable for 22mm cutouts. The bulb is replaceable and it uses a T3-1/4 wedge base bulb up to 250V. This pilot light assembly is commonly used on control panels and industrial equipment. With its durable construction, it provides long-lasting illumination to visually indicate power or status.