Learn More
Grainger Pilot Light Lens 30mm Plastic Amber
Catalog No. 502752800
Encompass_Preferred
The Eaton 14A158 is a 30mm pilot light lens for indicating the status of electrical equipment. It has a clear polycarbonate lens and chrome-plated brass housing suitable for 22mm cutouts. The bulb is replaceable and it uses a T3-1/4, 0.2A bulb. This pilot light can withstand temperatures up to 85°C. The overall dimensions are 30mm diameter x 35mm depth. This pilot light lens provides a visual status indicator for industrial controls and machinery.
Catalog No. 50-275-2800 Supplier Grainger Supplier No. 14A158
The Eaton 14A158 is a 30mm pilot light lens for indicating the status of electrical equipment. It has a clear polycarbonate lens and chrome-plated brass housing suitable for 22mm cutouts. The bulb is replaceable and it uses a T3-1/4, 0.2A bulb. This pilot light can withstand temperatures up to 85°C. The overall dimensions are 30mm diameter x 35mm depth. This pilot light lens provides a visual status indicator for industrial controls and machinery.