Learn More
Grainger Pilot Light Lens 30mm Plastic Green
Catalog No. 502752802
Encompass_Preferred
The Eaton 14A159 is a 30mm diameter pilot light lens for indicating the status of electrical equipment. It has a clear polycarbonate lens and chrome-plated brass housing suitable for 22mm cutout. The bulb utilizes a T3.5 bi-pin base, draws 150mA at 6-8V AC/DC, and provides 130mcd brightness. It mounts via a threaded ring and has an IP65 rating for dust and water resistance. The overall dimensions are 30mm diameter x 35mm depth. This pilot light lens allows clear indication in industrial controls and machinery.
Catalog No. 50-275-2802 Supplier Grainger Supplier No. 14A161
The Eaton 14A159 is a 30mm diameter pilot light lens for indicating the status of electrical equipment. It has a clear polycarbonate lens and chrome-plated brass housing suitable for 22mm cutout. The bulb utilizes a T3.5 bi-pin base, draws 150mA at 6-8V AC/DC, and provides 130mcd brightness. It mounts via a threaded ring and has an IP65 rating for dust and water resistance. The overall dimensions are 30mm diameter x 35mm depth. This pilot light lens allows clear indication in industrial controls and machinery.