Learn More
Grainger Pilot Light Lens 30mm Plastic Red
Catalog No. 502750380
Encompass_Preferred
The Eaton 14A162 is a 30mm diameter pilot light lens for indicating the status of electrical equipment. It has a clear polycarbonate lens and chrome-plated brass housing suitable for 22mm cutout. The bulb utilizes a T3.5 bi-pin base, draws 150mA at 6-8V AC/DC, and provides 130mcd brightness. It mounts via a threaded ring and has an IP40 rating against solid objects over 1mm. The overall dimensions are 30.5mm diameter x 35.3mm length. This pilot light lens allows clear indication in industrial control panels and machinery.
Catalog No. 50-275-0380 Supplier Grainger Supplier No. 14A162
The Eaton 14A162 is a 30mm diameter pilot light lens for indicating the status of electrical equipment. It has a clear polycarbonate lens and chrome-plated brass housing suitable for 22mm cutout. The bulb utilizes a T3.5 bi-pin base, draws 150mA at 6-8V AC/DC, and provides 130mcd brightness. It mounts via a threaded ring and has an IP40 rating against solid objects over 1mm. The overall dimensions are 30.5mm diameter x 35.3mm length. This pilot light lens allows clear indication in industrial control panels and machinery.