Learn More
MSC Tingley Pilot Knee Boot, Steel Toe, PVC, 15 in. Height, 1 Pair
Catalog No. 5000685422
Encompass
These 15 in. knee boots provide 100% waterproof protection and are designed for use in construction, agriculture, and industrial environments. Featuring injection-molded, seamless construction, these boots include a steel toe and puncture-resistant midsole to meet rigorous safety standards. The design incorporates a self-cleaning, cleated outsole for improved traction and an expandable calf relief area for enhanced comfort.
  • Meets ASTM F2413 M I/75 C/75 EH standards for impact, compression, and electrical hazard protection
  • ASTM F3445 compliant slip-resistant outsole
  • Puncture-resistant steel midsole
  • Flexible upper material that stays supple in cold temperatures
  • Removable, moisture-wicking EVA cushioned insole
  • Calf relief expansion allows the boot to expand up to 1.5 in. for extra comfort
  • Chemical resistant to certain acids, fats, oils, and hydrocarbons
Catalog No. 50-006-85422 Supplier MSC Supplier No. 39906631
These 15 in. knee boots provide 100% waterproof protection and are designed for use in construction, agriculture, and industrial environments. Featuring injection-molded, seamless construction, these boots include a steel toe and puncture-resistant midsole to meet rigorous safety standards. The design incorporates a self-cleaning, cleated outsole for improved traction and an expandable calf relief area for enhanced comfort.
  • Meets ASTM F2413 M I/75 C/75 EH standards for impact, compression, and electrical hazard protection
  • ASTM F3445 compliant slip-resistant outsole
  • Puncture-resistant steel midsole
  • Flexible upper material that stays supple in cold temperatures
  • Removable, moisture-wicking EVA cushioned insole
  • Calf relief expansion allows the boot to expand up to 1.5 in. for extra comfort
  • Chemical resistant to certain acids, fats, oils, and hydrocarbons