Learn More
Medchemexpress LLC PILR-beta Human HE 10ug | 10ug
SDP
Catalog No. 5000187179
Encompass_Preferred
PILR- is part of a family of paired receptors that play a critical regulatory role in the immune system PILR- functions as an activating receptor and is thought to participate in cell signaling by binding to ITAM-bearing adapter molecules suggesting its role in transmitting immune response signals PILR-beta Protein Human (HEK293 His) is the recombinant human-derived PILR-beta protein expressed by HEK293 with N-His labeled tag
Catalog No. 50-001-87179 Supplier Medchemexpress LLC Supplier No. HYP7746310UG
PILR- is part of a family of paired receptors that play a critical regulatory role in the immune system PILR- functions as an activating receptor and is thought to participate in cell signaling by binding to ITAM-bearing adapter molecules suggesting its role in transmitting immune response signals PILR-beta Protein Human (HEK293 His) is the recombinant human-derived PILR-beta protein expressed by HEK293 with N-His labeled tag