Learn More
Medchemexpress LLC PILR-beta Mouse HE 5ug | 5ug
SDP
Catalog No. 5000186445
Encompass_Preferred
The PILR- protein is a paired immune system modulator and is considered a signal-activated receptor that associates with cell surface ITAM adapters such as DAP12 It interacts with CD99 and may help natural killer cells recognize target cells and activate dendritic cells PILR-beta Protein Mouse (HEK293 Fc) is the recombinant mouse-derived PILR-beta protein expressed by HEK293 with C-hFc labeled tag
Catalog No. 50-001-86445 Supplier Medchemexpress LLC Supplier No. HYP771435UG
The PILR- protein is a paired immune system modulator and is considered a signal-activated receptor that associates with cell surface ITAM adapters such as DAP12 It interacts with CD99 and may help natural killer cells recognize target cells and activate dendritic cells PILR-beta Protein Mouse (HEK293 Fc) is the recombinant mouse-derived PILR-beta protein expressed by HEK293 with C-hFc labeled tag