Learn More
Aviva Systems Biology Corp PILRA Antibody - N-terminal region (ARP46947_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46947_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PILRA. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PILRA. Alternate names: FDF03ApplicationWB. Species reactivity: Human
Catalog No. 50-014-59439