Learn More
Medchemexpress LLC Pilsicainide | 88069-49-2 | MFCD00903769 | 99.9% | 308.8 g/mol | C17H25ClN2O | 50 MG
SDP
Catalog No. 5000212451
Encompass_Preferred
Pilsicainide hydrochloride is the hydrochloride salt of pilsicainide, an orally active sodium channel blocker and potent class Ic antiarrhythmic agent. It is supplied for research use in electrophysiology and cardiac pharmacology to investigate sodium channel modulation and arrhythmia mechanisms.
  • Orally active sodium channel blocker
  • Class Ic antiarrhythmic agent
  • Hydrochloride salt form for improved solubility
  • Reported purity approximately 99.9% for research use
  • Used in electrophysiology and cardiac pharmacology studies
  • Provided in small laboratory quantities for experimental work
Catalog No. 50-002-12451 Supplier Medchemexpress LLC Supplier No. HY10124550MG
Pilsicainide hydrochloride is the hydrochloride salt of pilsicainide, an orally active sodium channel blocker and potent class Ic antiarrhythmic agent. It is supplied for research use in electrophysiology and cardiac pharmacology to investigate sodium channel modulation and arrhythmia mechanisms.
  • Orally active sodium channel blocker
  • Class Ic antiarrhythmic agent
  • Hydrochloride salt form for improved solubility
  • Reported purity approximately 99.9% for research use
  • Used in electrophysiology and cardiac pharmacology studies
  • Provided in small laboratory quantities for experimental work