Learn More
Medchemexpress LLC Pilsicainide hydrochloride | 88069-49-2 | 99.9% | 308.85 | C17H25ClN2O | 5 MG
SDP
Catalog No. 5000212721
Encompass_Preferred
Pilsicainide hydrochloride is an orally active sodium channel blocker used in research on cardiac electrophysiology. It functions as a potent class Ic antiarrhythmic agent and is provided as a solid suitable for laboratory studies.
  • Orally active sodium channel blocker.
  • Potent class Ic antiarrhythmic agent.
  • High purity (99.94%).
  • White to off-white solid appearance.
  • Available in small-mass packages for analytical and pharmacological work.
  • Stable when stored under recommended conditions.
Catalog No. 50-002-12721 Supplier Medchemexpress LLC Supplier No. HY1012455MG
Pilsicainide hydrochloride is an orally active sodium channel blocker used in research on cardiac electrophysiology. It functions as a potent class Ic antiarrhythmic agent and is provided as a solid suitable for laboratory studies.
  • Orally active sodium channel blocker.
  • Potent class Ic antiarrhythmic agent.
  • High purity (99.94%).
  • White to off-white solid appearance.
  • Available in small-mass packages for analytical and pharmacological work.
  • Stable when stored under recommended conditions.