Learn More
Medchemexpress LLC Pilsicainide hydrochloride | 88069-49-2 | MFCD00903769 | 99.9% | 308.85 | C17H25ClN2O | 10 MG

Supplier: Medchemexpress LLC HY10124510MG
Pilsicainide hydrochloride is an orally active class Ic antiarrhythmic agent and potent sodium channel blocker used as a research reagent. Supplied as a high-purity hydrochloride salt for in vitro and in vivo pharmacology and electrophysiology studies.
- High purity: 99.9%.
- Molecular weight 308.85 g·mol-1.
- Chemical formula C17H25ClN2O.
- Soluble in water and DMSO; sonication recommended for dissolution.
- Available as a 10 MG pack for small-scale studies.
- Storage recommended at 4°C for solid; in solution store at -80°C up to 6 months.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.