Learn More
Medchemexpress LLC Pilsicainide hydrochloride | 88069-49-2 | MFCD00903769 | 99.9% | 308.85 g·mol⁻¹ | C17H25ClN2O | 25 MG
SDP
Catalog No. 5000211110
Encompass_Preferred
Pilsicainide hydrochloride is the hydrochloride salt of pilsicainide, an orally active sodium channel blocker and potent class Ic antiarrhythmic agent. It is supplied as a high-purity solid for preclinical pharmacology, electrophysiology, and biochemical research.
  • Orally active sodium channel blocker.
  • Potent class Ic antiarrhythmic activity.
  • High reported purity (>99.9%).
  • Molecular formula C17H25ClN2O; molecular weight 308.85 g·mol⁻¹.
  • Suitable for pharmacological and electrophysiological studies.
  • Available in research-scale pack sizes.
Catalog No. 50-002-11110 Supplier Medchemexpress LLC Supplier No. HY10124525MG
Pilsicainide hydrochloride is the hydrochloride salt of pilsicainide, an orally active sodium channel blocker and potent class Ic antiarrhythmic agent. It is supplied as a high-purity solid for preclinical pharmacology, electrophysiology, and biochemical research.
  • Orally active sodium channel blocker.
  • Potent class Ic antiarrhythmic activity.
  • High reported purity (>99.9%).
  • Molecular formula C17H25ClN2O; molecular weight 308.85 g·mol⁻¹.
  • Suitable for pharmacological and electrophysiological studies.
  • Available in research-scale pack sizes.