Learn More
Medchemexpress LLC PIM-447 (dihydrochloride) | 1820565-69-2 | 99.8% | 513.38 | 1 MG
SDP
Catalog No. 5000365171
Encompass_Preferred
PIM-447 dihydrochloride (LGH447 dihydrochloride) is a potent, orally available, and selective pan-PIM kinase inhibitor. With Ki values of 6, 18, and 9 pM for PIM1, PIM2, and PIM3, it displays dual antimyeloma and bone-protective effects, and induces apoptosis. This compound is for research use only.
  • Potent, orally available, and selective pan-PIM kinase inhibitor
  • Inhibits PIM1, PIM2, and PIM3 with picomolar affinity
  • Exhibits dual antimyeloma and bone-protective effects
  • Induces apoptosis in multiple myeloma cells
  • Increases annexin-V levels and promotes caspase cleavage
  • Controls tumor progression in mouse models
Catalog No. 50-003-65171 Supplier Medchemexpress LLC Supplier No. 1MGHY19322B
PIM-447 dihydrochloride (LGH447 dihydrochloride) is a potent, orally available, and selective pan-PIM kinase inhibitor. With Ki values of 6, 18, and 9 pM for PIM1, PIM2, and PIM3, it displays dual antimyeloma and bone-protective effects, and induces apoptosis. This compound is for research use only.
  • Potent, orally available, and selective pan-PIM kinase inhibitor
  • Inhibits PIM1, PIM2, and PIM3 with picomolar affinity
  • Exhibits dual antimyeloma and bone-protective effects
  • Induces apoptosis in multiple myeloma cells
  • Increases annexin-V levels and promotes caspase cleavage
  • Controls tumor progression in mouse models