Learn More
Medchemexpress LLC PIM-447 dihydrochloride | 1820565-69-2 | 99.8% | 513.38 | 1 ML
SDP
Catalog No. 5000385684
Encompass_Preferred
PIM447 dihydrochloride (LGH447 dihydrochloride) is a potent, orally available, and selective pan-PIM kinase inhibitor, with K¡ values of 6, 18, and 9 pM for PIM1, PIM2, and PIM3, respectively. PIM447 dihydrochloride displays dual antimyeloma and bone-protective effects, and induces apoptosis.
  • Potent, orally available, and selective pan-PIM kinase inhibitor.
  • K¡ values of 6, 18, and 9 pM for PIM1, PIM2, and PIM3, respectively.
  • Displays dual antimyeloma and bone-protective effects.
  • Induces apoptosis.
Catalog No. 50-003-85684 Supplier Medchemexpress LLC Supplier No. HY19322B10MM/1ML
PIM447 dihydrochloride (LGH447 dihydrochloride) is a potent, orally available, and selective pan-PIM kinase inhibitor, with K¡ values of 6, 18, and 9 pM for PIM1, PIM2, and PIM3, respectively. PIM447 dihydrochloride displays dual antimyeloma and bone-protective effects, and induces apoptosis.
  • Potent, orally available, and selective pan-PIM kinase inhibitor.
  • K¡ values of 6, 18, and 9 pM for PIM1, PIM2, and PIM3, respectively.
  • Displays dual antimyeloma and bone-protective effects.
  • Induces apoptosis.