Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PIM-447 dihydrochloride | 1820565-69-2 | MFCD30489747 | 99.3% | 513.38 | C24H25Cl2F3N4O | 10 MG
SDP

Supplier:  Medchemexpress LLC HY19322B10MG

Encompass_Preferred

PIM-447 dihydrochloride is the dihydrochloride salt of a potent, orally available pan-PIM kinase inhibitor used in preclinical research. It modulates apoptosis and JAK/STAT signaling and has been investigated for antimyeloma and bone-protective effects. This reagent is supplied for laboratory research use only.

  • Potent pan-PIM kinase inhibition with low picomolar Ki values.
  • Demonstrated antimyeloma and bone-protective activity in preclinical studies.
  • High solubility in water and DMSO (H2O: 50 mg/mL; DMSO: ≥46.7 mg/mL).
  • High purity (≈99.3%).
  • Provided as a 10 mg solid suitable for preparing concentrated stock solutions.
  • Stability supported by recommended storage conditions in solvent (-80°C 1 year; -20°C 6 months).
  • Intended for research use only; not for human or veterinary use.

Catalog No. 50-002-94720


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.