Learn More
TARGETMOL CHEMICALS INC PIM1/AKK1-IN-1 5MG
SDP
Catalog No. 502579816
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Pim1/AKK1-IN-1 (LKB1/AAK1 dual inhibitor) is a potent multi-kinase inhibitor with Kd values of 35 nM/53 nM/75 nM/380 nM for Pim1/AKK1/MST2/LKB1 respectively and also inhibits MPSK1 and TNIK. purity: 98%
Catalog No. 50-257-9816 Supplier TARGETMOL CHEMICALS INC Supplier No. T50935MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Pim1/AKK1-IN-1 (LKB1/AAK1 dual inhibitor) is a potent multi-kinase inhibitor with Kd values of 35 nM/53 nM/75 nM/380 nM for Pim1/AKK1/MST2/LKB1 respectively and also inhibits MPSK1 and TNIK. purity: 98%