Learn More
Aviva Systems Biology Corp PIM1 Antibody - N-terminal region (ARP56390_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56390_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PIM1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PIM1. Alternate names: PIMApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-61839