Learn More
Selleck Chemical LLC PIM447
SDP
Catalog No. 5000000478
Encompass_Preferred
PIM447 (LGH447) Hydrochloride is a novel pan-PIM kinase inhibitor with Ki values of 6 pM 18 pM 9 pM for PIM1 PIM2 PIM3 respectively It also inhibits GSK3 PKN1 and PKC but at a significantly lower potency with IC50 between 1 and 5 M ( 105-fold differential relative to the Ki on PIMs) PIM447 induces apoptosis
Catalog No. 50-000-00478 Supplier Selleck Chemical LLC Supplier No. S79855MG
PIM447 (LGH447) Hydrochloride is a novel pan-PIM kinase inhibitor with Ki values of 6 pM 18 pM 9 pM for PIM1 PIM2 PIM3 respectively It also inhibits GSK3 PKN1 and PKC but at a significantly lower potency with IC50 between 1 and 5 M ( 105-fold differential relative to the Ki on PIMs) PIM447 induces apoptosis